Learn More
HIC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22788-1-AP-20UL
 View more versions of this product
Product Code. 16802095
  • 145.00 EUR / 20µL
Description
HIC2 contains 5 C2H2-type zinc fingers and 1 BTB (POZ) domain. It belongs to the krueppel C2H2-type zinc-finger protein family, Hic subfamily and is a transcriptional repressor.
Specifications
Specifications
HIC2
Polyclonal
Unconjugated
HIC2
Hic 2, Hic 3, HIC2, HRG22, hypermethylated in cancer 2, KIAA1020, ZBTB30
Rabbit
Antigen Affinity Chromatography
RUO
23119
-20°C
Liquid
Immunoprecipitation, Western Blot
0.61 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96JB3
HIC2
HIC2 Fusion Protein Ag18789
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions