Learn More
Nabertherm™ High-Temperature Furnaces with Molybdenum Disilicide Heating Elements and Fiber Insulation
Shop All Nabertherm Products
Click to view available options
Capacity (Metric):
8 L
16 L
Temperature (Metric):
1600°C
1750°C
1800°C
Unit Size:
Each
Description
- Dual shell housing with fan cooling for low shell temperatures
- Heating from both sides via molybdenum disilicide heating elements
- High-quality fiber insulation backed by special insulation
- Side insulation constructed with tongue and groove blocks provides for low heat loss to the outside
- Long-life roof insulation with special suspension
- Chain-guided parallel swivel door for defined opening and closing of the door
- Vapor vent in the furnace roof with motorized exhaust air flaps, controlled via the extra function of the controller
- Heating elements switched via thyristors
Specifications
Specifications
| Controller Type | P570 |
| Capacity (Metric) | 8 L |
| Temperature (Metric) | 1750°C |
| Includes | Molybdenum Disilicide Heating Elements and Fiber Insulation |
| Dimensions (D x W x H) Interior | 300 x 150 x 150 mm |
| Dimensions (L x W x H) Exterior | 640 x 740 x 1755 mm |
| Voltage | 400 V |
| Wattage | 8500 W |