missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen Capto Blue

Excellent for method optimization and parameter screening due to the 10 cm bed height

Brand:  Cytiva 28-9924-74


Product Code. 11340132

  • 217.00 EUR / Each
Estimated Shipment: 18-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Prepacked with Capto Blue
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume for fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same flow velocity
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HiScreen Capto Blue
7.7 mm
3 bar [0.3 MPa] (44 psi)
Affinity Column
4.7 mL
1.2 to 2.3 mL/min.
2° to 8°C
Group specific affinity
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiScreen Capto Blue, 1 x 4.7 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers