missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ Sepharose™ Prepacked Fast Flow HIC Columns

Designed for method optimization and parameter screening

158.00 EUR

Specifications

Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
Height (English) 3.93 in.
View More Specs

Products 4
Product Code Brand Stationary Phase Price Quantity & Availability  
Product Code Brand Stationary Phase Price Quantity & Availability  
10818910
View Documents
Cytiva
28-9269-84
Butyl Sepharose
158.00 EUR
Each
In Stock
to see stock.
Add to basket
 
10640771
View Documents
Cytiva
28-9269-85
Butyl-S-Sepharose
158.00 EUR
Each
In Stock
to see stock.
Add to basket
 
10828910
View Documents
Cytiva
28-9269-89
Phenyl (low sub) Sepharose
158.00 EUR
Each
In Stock
to see stock.
Add to basket
 
10871601
View Documents
Cytiva
28-9269-86
Octyl Sepharose
158.00 EUR
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • Ready-to-use columns for convenient process development optimization
  • Standard aliphatic/aromatic Hydrophobic Interaction Chromatography media for capture and intermediate purification
  • Bed height of 10cm designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume of 4.7mL for fast result and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with same chromatography media using same linear fluid velocity

  • Bed dimension: 7.7 × 100mm
  • Matrix: 4% cross-linked agarose/6% cross-linked agarose
  • pH stability working range (media): 3 to 13
  • Average particle size: 90μm
  • Material (column hardware and column tube): polypropylene
  • Column I.D.: 7.7mm
  • Pressure max.: 1.5 bar (22psi)
  • Flow velocity: <240/450cm/hr.

Specifications

Specifications

4.7mL
7.7mm
3.93 in.
Polypropylene
3 to 13
4° to 30°C
Convenient process development optimization parameter screening
0.3 in.
1 x 4.7mL
100mm
1.5 bar
45 to 165μm
Ion Exchange Column
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiScreen Butyl FF

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.