missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HisPur™ Cobalt Spin Columns, 3 mL

Gently purify His-tagged recombinant fusion proteins using cobalt (Co2+) immobilized metal chelate affinity (IMAC) beads, spin columns and plates.

Brand:  Thermo Scientific™ 89969

178.21 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 11864181

  • 251.00 EUR / Pack of 5
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ HisPur Cobalt Resin is an immobilized metal chelate affinity (IMAC) support for specific, fast and gentle His-tagged purification.

Additional Information and Online Ordering

HisPur Cobalt Resins, Columns and Kits use a tetradentate chelating resin charged with divalent cobalt (Co2+)to enable specific, fast and gentle purification of histidine-tagged fusion proteins from a variety of sample types. HisPur Cobalt Resin is available in several package sizes of resin slurry, three sizes of centrifuge-ready columns, complete purification kits, two sizes of FPLC-ready chromatography cartridges, and 96-well filter plates for high throughput needs. With these options, high-quality His-tag protein purification is possible at nearly any scale. HisPur Cobalt Resin is superior to traditional nickel-based immobilized metal affinity chromatography (IMAC) systems. Compared to Ni-IDA and tetradentate Ni2+chelate resins, HisPur Resin binds histidine-tagged proteins with higher specificity (less off-target binding) and releases them with gentler conditions (lower concentrations of imidazole). Use HisPur Columns and Cartridges to obtain high-purity His-tagged proteins with no metal contamination.

Highlights:

  • High purity – obtain more than 10 milligrams of pure His-tagged protein per milliliter of resin without optimizing imidazole washing conditions

  • Low metal leaching – no metal contamination in eluted histidine-tagged protein sample

  • Versatility – purify proteins under native or denaturing conditions; compatible with cell lysis reagents and a variety of buffer additives

  • Flexibility – available as bulk resin or predispensed columns, chromatography columns and 96-well filter plates

  • Cost effective – resin can be reused several times or discarded after single use

  • Superior – performs better than other commercially available IMAC Resins

Available Formats:

  • Resin Slurries – crosslinked 6% beaded agarose; 10mL, 100mL, 500mL bottles
  • Spin Columns – 0.2mL (microcentrifuge), 1mL, 3mL columns
  • Purification Kits – complete kits in all three column sizes
  • Chromatography Cartridges – compatible with automated liquid chromatography systems and with syringe processing
  • 96-well Spin Plates – pre-dispensed 96-well filter plates, compatible with centrifugation and vacuum manifold systems for manual or automated purification. The plates enable fast, consistent well-to-well and plate-to-plate reproducibility for small-scale, high-throughput separations
  • Bulk and Custom – let us know your needs for bulk HisPur Cobalt Resin

Includes:

Kits contain HisPur Cobalt Columns, equilibration/wash buffer and elution buffer

TRUSTED_SUSTAINABILITY
Specifications

Specifications

3 mL
Store at 4°C
HisPur™
Recombinant
Spin Column
Spin Column
HisPur Cobalt Spin Columns, 3mL
Liquid Suspension
5 Columns
His-Tagged Protein
Metal Chelate, Divalent Cobalt
Affinity, Agarose Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.