missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-ABC Monoclonal Antibody (W6/32), NovaFluor™ Blue 510, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
155.00 EUR - 344.00 EUR
Specifications
Antigen | HLA-ABC |
---|---|
Clone | W6/32 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
17826537
|
Invitrogen
H020T03B01 |
100 Tests |
344.00 EUR
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17896697
|
Invitrogen
H020T02B01 |
25 Tests |
155.00 EUR
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA-class I major histocompatibility (MHC) antigens are intrinsic membrane glycoproteins expressed on nucleated cells and noncovalently associated with an invariant beta2 microglobulin. They carry foreign determinants important for immune recognition by cytotoxic T cells, thus important for anti-viral and anti-tumor defense. Human HLA-class I antigens are represented by HLA-A, HLA-B and HLA-C molecules. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Spécification
HLA-ABC | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide, pH 7.2 | |
A29 protein, alpha chain of MHC class II, antigen presenting molecule, AS, Aw-33, Aw-74, B-4901, B-5001, Bw-47, Bw-50, class I MHC, HLA-B antigen, D6S204, epididymis secretory protein Li 83, H-2K, H2-K, H-2K(d), HEL-S-83, histocompatibility antigen, HLA A, HLA -B, HLA B-40011, HLA B44, HLA C, HLA class I antigen, HLA class I antigen HLA-B, HLA class I histocompatibility antigen C alpha chain, HLA class I histocompatibility antigen Cw-1 alpha chain, HLA class I histocompatibility antigen, A alpha chain, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-11 alpha chain-like, HLA class I histocompatibility antigen, A-30 alpha chain, HLA class I histocompatibility antigen, A-33 alpha chain, HLA class I histocompatibility antigen, A-74 alpha chain, HLA class I histocompatibility antigen, B alpha chain, HLA class I histocompatibility antigen, B-47 alpha chain, HLA class I histocompatibility antigen, B-50 alpha chain, HLA class I histocompatibil | |
HLA-A, HLA-B, HLA-C | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
W6/32 | |
Flow Cytometry | |
NovaFluor Blue 510 | |
Mouse | |
Human | |
P01889, P04439, P10321 | |
3105, 3106, 3107 | |
IgG2a κ | |
Affinity chromatography | |
HLA-C |