missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DR/DP Antibody (CL10961), Novus Biologicals™
Mouse Monoclonal Antibody
378.00 EUR - 618.00 EUR
Specifications
Antigen | HLA DR/DP |
---|---|
Clone | CL10961 |
Dilution | Western Blot 1 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18625694
|
Novus Biologicals
NBP3-21186-25ul |
25 μg |
378.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18673285
|
Novus Biologicals
NBP3-21186-100ul |
100 μg |
618.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA DR/DP Monoclonal antibody specifically detects HLA DR/DP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
HLA DR/DP | |
Western Blot 1 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
This antibody was developed against a Synthetic Peptide of P01903. Actual immunogen sequence is proprietary. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL10961 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology | |
PBS, pH 7.2, 40% glycerol | |
3122 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HLA DR/DP Antibody (CL10961), Novus Biologicals™