Learn More
Macherey-Nagel™ Holder For NANOCOLOR Test Tubes
The NANOCOLOR test tube holder accommodates 15 round glass tubes (16 mm OD) and 2 tubes for sample digestion (22 mm OD).
Brand:  Macherey-Nagel™ 91623
Product Code. 16237730
  • 81.85 EUR / Each
Description
  • Tube holder
  • Holds 15 tubes (16 mm OD) and 2 sample digestion tubes (22 mm OD) simultaneously
Specifications
Specifications
Tube Rack
15 Tubes (16 mm OD) and 2 Sample Digestion Tubes (22 mm OD) Simultaneously