Learn More
Macherey-Nagel™ Holder For NANOCOLOR Test Tubes
The NANOCOLOR test tube holder accommodates 15 round glass tubes (16 mm OD) and 2 tubes for sample digestion (22 mm OD).
Brand: Macherey-Nagel™ 91623
This item is not returnable.
View return policy
Description
- Tube holder
- Holds 15 tubes (16 mm OD) and 2 sample digestion tubes (22 mm OD) simultaneously
Specifications
Tube Rack | |
15 Tubes (16 mm OD) and 2 Sample Digestion Tubes (22 mm OD) Simultaneously |