Learn More
Thermo Scientific™ Hollow Cathode Lamps for iCE™ 3000 Series Atomic Absorption Spectrometers
For maximum flexibility from your Thermo Scientific™ iCE™ 3000 Series AAS system, we offer a huge range of pre-coded and un-coded Hollow Cathode Lamps.
Brand: Thermo Scientific™ 942339321281
Description
For maximum flexibility from your Thermo Scientific™ iCE™ 3000 Series Atomic Absorption spectrometer, a huge range of pre-coded and un-coded Thermo Scientific™ Hollow Cathode Lamps are available, to handle your chosen elements for analysis. Whether you require a single dedicated element or a multi-element cathode lamp, there is a one to match your needs, whether coded or un-coded.
For maximum flexibility from your Thermo Scientific™ iCE™ 3000 Series Atomic Absorption spectrometer, a huge range of pre-coded and un-coded Thermo Scientific Hollow Cathode Lamps are available, to handle your chosen elements for analysis. Whether you require a single dedicated element or a multi-element cathode lamp, there is a one to match your needs, whether coded or un-coded.Specifications
Hollow Cathode Lamps, Uncoded | |
iCE 3000 Series Atomic Absorption Spectrometers | |
Co/Cu/Fe Uncoded |