Learn More
Sperian™ Honeywell™ Clearways Faceshield
10.03 EUR valid until 2024-05-31
Use promo code "21638" to get your promotional price.
Use promo code "21638" to get your promotional price.
Click to view available options
Product Type:
Browguard with elastic headband
Visor
Lens Coating:
Anti-fog
Uncoated
Lens Material:
Acetate
N/A
Polycarbonate
Description
- Recommended for the agriculture, chemical and forestry industries
- Protects against hazards including abrasion, airborne particles, biohazards, chemicals, impacts, splashes and more
- Safe, practical, economical integral visor fitting ensures extra protection
Order Info
Note: The image shows both the Browguard and the Visor. Both pieces must be ordered separately. See Browguard (11886980), and Visors (11806990, 12652733, or 11896980).
Specifications
Certifications/Compliance | EN 166 1 F |
Lens Coating | Uncoated |
Product Type | Browguard with elastic headband |
Model | CB14 |
No. per Pack | 1 Ea. |
Material | Polypropylene |
Lens Material | N/A |