Learn More
Sperian™ Honeywell™ MAXLITE™ LS-400 Earplugs Refill
Click to view available options
:
Color: Green
Description
- Low-pressure foam expands gently inside the ear canal for comfortable long-term wear
- Eliminates feeling of “tightness”
- Ideal for process industries with low tolerances for packaging waste
Order Info
dimProductType
Specifications
Specifications
| Color | Green |
| Size | Universal |
| Quantity | 200/Pk. |
| Plug Shape | T-Shape |
| Product Type | Earplug Refill |
| Disposable | Single-use |