Learn More
Honeywell™ V-Maxx Safety Goggles
missing translation for 'orderingAttributeHoverText'
:
Clear FogBan Lens
Descrizione
- Sleek wrap-around styling for a 180°, clear field of vision
- Adjustable elastic or neoprene headband
- Peel-off goggle covers available to extend the life of the lenses
- Recommneded for the chemical, construction, manufacturing and welding industries
- Protects against hazards such as abrasion, airborne particles, biohazards and chemicals, among many others
Specifica
Specifica
| Lens Coating | Anti-fog |
| Lens Tint | Clear |
| Product Type | Safety Spectacles |
| For Use With (Application) | Industry Automotive and Part Manufacturer Building and Construction Food Industries Industrial Cleaning Laboratory Maintenance |
| Frame Color | Clear |
| No. per Pack | 1 Ea. |
| Material | Plastic |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto