missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ HPLC Air Valve, for PFAS Analytic
GREENER_CHOICE

Product Code. 18606498 Shop All SCAT Europe Products
Click to view available options
Quantity:
1 Ea.
10 Pk.
8 Pk.
Unit Size:
1 sheet
10 sets
8 sets
This item is not returnable. View return policy

Product Code. 18606498

Brand: SCAT™ 397010PP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The SCAT air valve blocks harmful vapours and ensures safe pressure equalisation in the storage tanks of your HPLC. Solvent mixtures remain stable: thanks to closed system without evaporation.

  • With Luer-Lock connection for gassing the solvents and change indicator for timely change.
  • Made of PP for PFAS analysis of polyfluorinated chemicals (formerly: PFC)
  • UNF 1/4 in. 28G thread size fits to any SCA PFAS-Analytic Safety Cap for PFAS Analysis

Specifications

For Use With (Equipment) SCAT Safety Caps for PFAS-Analytic
Material PP
Thread Size UNF 1/4 in. 28G
For Use With (Application) HPLC analysis
Type Valve
Quantity 10 Pk.
Color Red, Green
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.