missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsp47 Mouse anti-Human, Clone: 950811, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB91661-SP
This item is not returnable.
View return policy
Description
Hsp47 Monoclonal specifically detects Hsp47 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
Hsp47 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
(collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, BERF-1, CBP1, CBP2, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), OI10, PPROM, proliferation-inducing gene 14, RA-A47, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH1, SERPINH2colligin | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
871 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
950811 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
P50454 | |
SERPINH1 | |
Chinese hamster ovary cell line CHO-derived recombinant human HSP47 Ala19-Asp412 Accession # P50454 | |
25 μg | |
Primary | |
Detects human HSP47 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with mouse HSP47 is observed. | |
Human | |
Purified |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction