missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HspB2 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
193.30 EUR - 457.87 EUR
Specifications
Antigen | HspB2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18331716
|
Bio-Techne
NBP3-04483-20UL |
20 μL |
193.30 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18396096
|
Bio-Techne
NBP3-04483-100UL |
100 μL |
457.87 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HspB2 Polyclonal specifically detects HspB2 in Mouse, Rat samples. It is validated for Western Blot.Specifications
HspB2 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
3316 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
DMPK-binding protein, HspB2, MKBP | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-182 of human HSPB2 (NP_001532.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |