missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ACE2 (aa1-615) Fc Chimera Recombinant Protein
HEK293 cells Recombinant Protein
Specifications
Accession Number | Q9BYF1 |
---|---|
Concentration | 1 mg/mL |
For Use With (Application) | Control, ELISA, Western Blot |
Formulation | PBS with no preservative |
Gene ID (Entrez) | 59272 |
Description
The protein encoded by this gene is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The external domain mediates cell attachment to the extracellular matrix molecule vitronectin, while its intracellular domain interacts with the dynein light chain Tctex-1/DYNLT1. The gene is specific to the primate lineage, and serves as a cellular receptor for poliovirus in the first step of poliovirus replication. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
Q9BYF1 | |
Control, ELISA, Western Blot | |
59272 | |
HEK293 cells | |
RUO | |
ACE2 | |
Unconjugated | |
Lyophilized |
1 mg/mL | |
PBS with no preservative | |
Human ACE2 (aa1-615) Fc Chimera | |
-20°Cor to +80°C if preferred | |
2010305L05Rik; ACE 2; Ace2; ACEH; ACE-related carboxypeptidase; angiotensin I converting enzyme (peptidyl-dipeptidase A) 2; angiotensin I converting enzyme 2; angiotensin-converting enzyme 2; Angiotensin-converting enzyme homolog; anigotensin I converting enzyme 2; anigotensin-converting enzyme-related carboxypeptidase; metalloprotease MPROT15; peptidyl-dipeptidase A; Processed angiotensin-converting enzyme 2; renal angiotensin-converting enzyme 2; UNQ868/PRO1885 | |
Human | |
Recombinant | |
>95% |