Learn More
Novus Biologicals™ Human Artery Whole Tissue Lysate (Adult Whole Normal)
Brand:  Novus Biologicals™ NB820-59175
Product Code. 18253223
  • 533.00 EUR / 1mg
Description
The Human Artery Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Western Blot
1 mg
Total Protein - Human Adult Normal Tissue: Blood Vessel: Artery
Normal
Human
Human Artery Whole Tissue Lysate (Adult Normal)
Adult
Vascular
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Human
5 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native
For Research Use Only