missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human BD-2 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 300-49-250UG
This item is not returnable.
View return policy
Description
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The alpha-defensins are distinguished from the beta-defensins by the pairing of their three disulfide bonds. To date, six human beta-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. beta-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The beta-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. Beta-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds.Specifications
O15263 | |
protein with no preservative | |
4.3 | |
250 μg | |
-20°C | |
<1 EU/μg | |
DEFB4A | |
Unconjugated | |
GIGDPVTCLK SGAICHPVFC PRRYKQIGTC GLPGTKCCKK P | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
1673 | |
Human BD-2 | |
E. coli | |
RUO | |
BD-2; beta defensin 2; beta defensin-2; Beta-defensin 2; Beta-defensin 4 A; DEFB102; DEFB2; DEFB-2; DEFB4; DEFB4A; DEFB4B; Defensin beta 2; defensin beta 4 A; Defensin beta2; defensin, beta 2; defensin, beta 4; defensin, beta 4 A; Defensin-beta2; HBD-2; SAP1; Skin-antimicrobial peptide 1 | |
Human | |
Recombinant | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction