missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human BD-5, Animal-Free Recombinant Protein, PeproTech®
Recombinant Protein
103.00 EUR - 1174.00 EUR
Specifications
Accession Number | Q8NG35 |
---|---|
For Use With (Application) | Functional Assay |
Format | Lyophilized |
Formulation | protein with no preservative |
Gene ID (Entrez) | 245908, 504180 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17846418
|
Gibco™
AF-300-68-5UG |
5 μg |
103.00 EUR
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17886448
|
Gibco™
AF-300-68-250UG |
250 μg |
1174.00 EUR
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The alpha-defensins are distinguished from the beta-defensins by the pairing of their three disulfide bonds. To date, six human beta-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. beta-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The beta-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. Beta-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds.Specifications
Q8NG35 | |
Lyophilized | |
245908, 504180 | |
DEFB105A | |
-20°C | |
<0.1 EU/ μg | |
Protein | |
Unconjugated | |
GLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR I | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
protein with no preservative | |
5.8 kDa | |
HPLC, SDS-PAGE | |
RUO | |
DEFB105A, DEFB105B | |
Human | |
Recombinant | |
E. coli |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title