missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BLMH ELISA Kit
Human BLMH quantitates human BLMH in serum, plasma, supernatant.
Brand: Invitrogen™ EH46RBX5
This item is not returnable.
View return policy
Description
Human BLMH quantitates human BLMH in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human BLMH.
The normal physiological role of BLM hydrolase is unknown, but it catalyzes the inactivation of the antitumor drug BLM (a glycopeptide) by hydrolyzing the carboxamide bond of its B-aminoalaninamide moiety thus protecting normal and malignant cells from BLM toxicity.Specifications
AI035728; BH; bleomycin hydrolase; BLM hydrolase; BLMH; BMH | |
0.085 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
BLMH | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
0.085 to 20 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q13867 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
5 x 96 Tests | |
Serum, 50 μL; Plasma, 50 μL; Supernatant, 50 μL | |
BLMH | |
4 hr 45 min |