Learn More
Novus Biologicals™ Human Brain Cerebral Cortex Membrane Tissue Lysate (Adult Membrane Normal)
Brand: Novus Biologicals™ NB820-60574
This item is not returnable.
View return policy
Description
The Human Brain Cerebral Cortex Membrane Tissue Lysate (Adult Membrane Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.Specifications
Western Blot | |
0.1 mg | |
Membrane Protein - Human Adult Normal Tissue: Brain: Cerebral Cortex | |
Normal | |
Human | |
Human Brain Cerebral Cortex Membrane Tissue Lysate (Adult Normal) | |
Adult |
Nervous | |
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
Membrane | |
Human | |
2 mg/mL | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
Product Content Correction
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction