Learn More
R&D Systems™ Human CD161 (NP_002249) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand:  R&D Systems™ RDC1672
Product Code. 18702799
  • 322.00 EUR / 10µg
Description
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
The Human CD161 (NP_002249) VersaClone cDNA clone contains the complete ORF for CD161, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CD161 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifications
Specifications
CD161
10 μg at 0.2 μg/μL
691bp
CD161, CD161 antigen, CLEC5B, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, KLRB1, Ly59, Natural killer cell surface protein P1A, NKR, NKRP1, NKR-P1, NKRP1A, NKR-P1AMGC138614, NKRP1ANKR
Human
NP_002249
cDNA is provided in 10mM Tris-Cl, pH 8.333
3820
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Special Offers
For Reasearch Use Only. Not for use in Humans.