missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dkk-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB10962-SP
This item is not returnable.
View return policy
Description
Dkk-1 Monoclonal specifically detects Dkk-1 in Human samples. It is validated for Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| Dkk-1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| O94907 | |
| DKK1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Dkk-1 Thr32-His266 Accession # O94907 | |
| 25 μg | |
| Primary | |
| Detects human Dkk-1 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Dkk-4, rhKremen-1, rhKremen-2 or rhLRP-6 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry, ELISA | |
| 141135 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, Dkk1, DKK-1, hDkk-1, SKdickkopf-1 like | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 22943 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction