Learn More
Proteintech™ Human FN1 ELISA Kit
Product Code. 16801466 Shop All Proteintech Products
Click to view available options
Target Species:
Human
Unit Size:
96 tests
Product Code. 16801466
Brand: Proteintech™ KE0003996T
Solid phase sandwich ELISA used to detect and quantify protein levels of endogenous FN1
  • Recognizes human FN1
  • Fibronectin 1 (FN1) is a high molecular weight glycoprotein, existing in soluble form in plasma (plasma FN1) and other body fluids and in insoluble form in extracellular matrix (cellular FN1)
    • Plasma FN1 (dimeric form) secreted by hepatocytes
    • Cellular FN (dimeric or cross-linked multimeric forms) made by fibroblasts, epitherlia and other cell types, deposited as fibrils in extracellular matrix
    • FN1 binds to cell surfaces through integrins and components (collagen, fibrin, heparin)
    • Involved in cell adhesion and migration processes (embryogenesis, wound healing, hemostasis, host defense, metastasis)
Process:
  • Antibody specific for FN1 pre-coated onto microwells
  • FN1 protein in samples captured by the coated antibody after incubation
  • After extensive washing, another antibody specific for FN1 is added to detect captured FN1 protein
  • For signal development, horseradish peroxidase (HRP)-conjugated antibody is added, followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm
Specifications
Accession Number NP_001293058
Assay Range 125 to 8000 pg/mL
Assay Sensitivity 47.2 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Human
Gene ID (Entrez) 2335
Host Species Human
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 3.5 hr.
UniProt ID P02751
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human VCAM-1/CD106 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine