Learn More
R&D Systems™ Human IFN-kappa (AAF67468.1) VersaClone cDNA
Brand: R&D Systems™ RDC3654
This item is not returnable.
View return policy
Specifications
IFN-kappa | |
56832 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
AAF67468.1 | |
IFNK, IFNkappa, IFN-kappa | |
10 μg |