missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-13 R alpha 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB6141-500
This item is not returnable.
View return policy
Description
IL-13R alpha 2 Monoclonal specifically detects IL-13R alpha 2 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| IL-13R alpha 2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cancer/testis antigen 19, CD213a2, CD213a2 antigen, CT19, IL-13 receptor subunit alpha-2, IL13BP, IL13R, IL-13R, IL13R alpha 2, IL-13R subunit alpha-2, IL13RA2, IL-13RA2, IL-13R-alpha-2, interleukin 13 binding protein, interleukin 13 receptor alpha 2 chain, interleukin 13 receptor, alpha 2, interleukin-13 receptor subunit alpha-2, Interleukin-13-binding protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3598 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 83807 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q14627 | |
| IL13RA2 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-13 R alpha 2 Cys22-Leu342 Accession # Q14627 | |
| 500 μg | |
| Primary | |
| Detects human IL-13 R alpha 2 in ELISAs. In sandwich ELISAs, no cross-reactivity or interference was observed with recombinant human (rh) IL-4 R, rhIL-5 R alpha , rhIL-9 R, rhIL-13, rhIL-13 R alpha 1, recombinant mouse (rm) IL-13, rmIL-13 R alpha 1, or rmIL-13 R alpha 2. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction