missing translation for 'onlineSavingsMsg'
Learn More

Human IL-6R alpha Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

210.00 EUR - 1775.00 EUR

Specifications

Antigen IL-6R alpha
Clone 556704R
Dilution Immunocytochemistry 3-25 ug/mL
Applications Immunocytochemistry
Classification Monoclonal
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18752679
GREENER_CHOICE
View Documents
R&D Systems
MAB2271R-SP
25 μg
210.00 EUR
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18730387
GREENER_CHOICE
View Documents
R&D Systems
MAB2271R-100
100 μg
440.00 EUR
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18120351
View Documents
R&D Systems
MAB2271R-500
500 ug
1775.00 EUR
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18179060
View Documents
R&D Systems
MAB2271R-01M
1 mg
1595.00 EUR
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-6R alpha Monoclonal specifically detects IL-6R alpha in Human samples. It is validated for Immunocytochemistry.
Specifications

Specifications

IL-6R alpha
Immunocytochemistry 3-25 ug/mL
Monoclonal
Purified
RUO
P08887
3570
IgG1
Protein A or G purified from cell culture supernatant
Detects human IL-6 R alpha in direct ELISAs.
556704R
Immunocytochemistry
Unconjugated
Mouse
Human
CD126, gp80, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor
IL6R
Primary
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.