missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Irisin/FNDC5 DuoSet ELISA
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Brand: R&D Systems™ DY9420-05
374.40 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
The Human Irisin/FNDC5 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Irisin/FNDC5.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
250.00 - 8000 pg/mL | |
ELISA | |
Human | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Solid Phase Sandwich ELISA | |
252995 | |
1 Kit (5 Plates) | |
Irisin/FNDC5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction