Learn More
Human/Mouse Cleaved Caspase-3 (Asp175) Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody has been used in 52 publications
Brand:  R&D Systems MAB835
Product Code. 18071862
  • 541.00 EUR / 50µg
Description
Caspase-3 Monoclonal specifically detects Caspase-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Caspase-3
Monoclonal
LYOPH
Western Blot 0.5 ug/mL, Immunohistochemistry 0.3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
U26943
CASP3
KLH-conjugated human Caspase-3 synthetic peptide CRGTELDCGIETD Accession # U26943
50 μg
Primary
Detects human and mouse Caspase-3 cleaved at Asp175. No cross-reactivity was detected with the full-length procaspase-3 or other caspases.
Human, Mouse
Purified
Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
269518
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama
Rabbit
Protein A or G purified from hybridoma culture supernatant
RUO
836
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions