missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat FKBP12.6 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF4174-SP
This item is not returnable.
View return policy
Description
FKBP12.6 Polyclonal specifically detects FKBP12.6 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| FKBP12.6 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 12.6 kDa FK506-binding protein, calstabin 2,12.6 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1B, 12.6 kDa, FK506-binding protein 12.6, FK506-binding protein 1B, FK506-binding protein 1B (12.6 kD), FKBP-12.6, FKBP12.6h-FKBP-12, FKBP1B, FKBP-1B, FKBP1L, FKBP9, Immunophilin FKBP12.6, OTK4, OTK4PPIase FKBP1B, peptidyl-prolyl cis-trans isomerase FKBP1B, PKBP1L, PPIase, Rotamase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2281 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 50 ug/mL | |
| P68106 | |
| FKBP1B | |
| E. coli-derived recombinant human FKBP12.6 Met1-Glu108 Accession # P68106 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat FKBP12.6 in Western blots. In Western blots, approximately 15% cross-reactivity with recombinant human FKBP12 and less than 1% cross-reactivity with recombinant human FKBP13 and FKBP25 is observed. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction