missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Osteopontin/OPN Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 22 publications
Brand: R&D Systems AF1433-SP
This item is not returnable.
View return policy
Description
Osteopontin/OPN Polyclonal specifically detects Osteopontin/OPN in Human samples. It is validated for ELISA, Immunohistochemistry, Neutralization, Immunocytochemistry.
Specifications
| Osteopontin/OPN | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SPP1 | |
| Human milk-derived Osteopontin/OPN | |
| 25 μg | |
| Primary | |
| Detects human Osteopontin/OPN in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| ELISA, Immunohistochemistry 5-15 ug/mL, Neutralization 0.300 - 3.60 ug/mL, Immunocytochemistry 1-15 ug/mL | |
| BNSP, Bone sialoprotein 1, Eta-1, MGC110940, Nephropontin, OPN, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), Spp1, SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6696 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction