Learn More
R&D Systems™ Human PIWIL1/HIWI (NP_004755) VersaClone cDNA
Brand:  R&D Systems™ RDC2832
Product Code. 18726868
  • 719.00 EUR / 10µg
Specifications
NP_004755
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
9271