missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Semaphorin 6B ELISA Kit

Product Code. 17155189
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17155189

Brand: Invitrogen™ EH408RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human Semaphorin 6B quantitates human Semaphorin 6B in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Semaphorin 6B.

May play a role in both peripheral and central nervous system development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Sema; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6 B; sema N; Sema VIB; Sema Z; SEMA6B; SEMAN; semaphorin 6 B; semaphorin VIB; semaphorin Z; Semaphorin-6 B; semaphorin-6 Ba; semaphorin-N; Semaphorin-Z; SEMA-VIB; SEMAZ; SEM-SEMA-Y; UNQ1907/PRO4353; VIb
Assay Range 0.6 to 150 ng/mL
Assay Sensitivity 0.6 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 10501
Gene Symbol SEMA6B
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target SEMA6B
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.