Learn More
Novus Biologicals™ Human Small Intestine Membrane Tissue Lysate (Adult Membrane Tumor)
Brand: Novus Biologicals™ NBL1-26436
This item is not returnable.
View return policy
Description
The Human Small Intestine Membrane Tissue Lysate (Adult Membrane Tumor) is derived from human.Specifications
Digestive/Waste | |
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors | |
Membrane | |
Human | |
2 mg/mL | |
-80°C. Avoid freeze-thaw cycles. | |
Native |
0.1 mg | |
Membrane Protein - Human Tumor Tissue: Small Intestine | |
Tumor | |
Human | |
Human Small Intestine Membrane Tissue Lysate (Adult Tumor) | |
Adult |