Learn More
BD Human Soluble Protein Master Buffer Kit
Click to view available options
No. of Tests:
100
500
Includes:
1 Bottle 30mL assay diluent, 1 bottle 5mL capture bead diluent, 1 bottle 5mL detection reagent diluent, 1 bottle 5mL capture bead diluent for Serum/Plasma, 1 bottle 130mL wash Buffer, 1 vial 0.25 mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9, 1 vial 1.0mL instrument setup bead F1, 1 vial 0.25mL instrument setup bead F9, 1 vial, 0.25mL PE instrument setup bead F1, 1 vial 0.5mL PE positive control detector
1 bottle 150mL assay diluent, 1 bottle 30mL capture bead diluent, 1 bottle 30mL detection reagent diluent, 1 bottle 30mL capture bead diluent for Serum/Plasma, 1 bottle 650mL wash buffer, 1 vial 0.25 mL instrument setup bead A1, 1 vial 0.25mL instrument
missing translation for 'unitSize'
1 set
Description
Includes
- Assay Diluent
- Capture Bead Diluent
- Detection Reagent Diluent
- Capture Bead Diluent for Serum/Plasma
- Wash Buffer
- Instrument Setup Bead A1
- Instrument Setup Bead A9
- Instrument Setup Bead F1
- Instrument Setup Bead F9
- PE Instrument Setup Bead F1
- PE Positive Control Detector
Buffers and instrument setup reagents provided in this kit have been optimized for analysis of analytes in tissue culture supernatants, plasma, and serum samples.
- Provides sufficient reagents for the quantitative analysis of 100 samples and 10 instrument setup procedures or 500 samples and 10 instrument setup procedures
- Has been optimized for use with the CBA Human Soluble Protein Flex Sets
- Working assay range for most analytes in this system is 10 to 2,500pg/mL
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Includes | 1 bottle 150mL assay diluent, 1 bottle 30mL capture bead diluent, 1 bottle 30mL detection reagent diluent, 1 bottle 30mL capture bead diluent for Serum/Plasma, 1 bottle 650mL wash buffer, 1 vial 0.25 mL instrument setup bead A1, 1 vial 0.25mL instrument |
| No. of Tests | 500 |
| Validated Application | RUO |
| Product Type | Cytometric Bead Array (CBA) Assay |