missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TAF9B (aa 135-157) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60968 (PA5-60968. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The protein encoded by this gene is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine. The encoded protein is involved in myelopoiesis, and defects in this gene have been associated with several myeloproliferative disorders. Two variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Accession Number | Q9HBM6 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 51616 |
| Name | Human TAF9B (aa 135-157) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | BC066223; Dn7; DN-7; neuronal cell death-related gene in neuron 7; neuronal cell death-related gene in neuron-7; neuronal cell death-related protein 7; Taf9b; TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31 kDa; TAF9L; TAF9-like RNA polymerase II TATA box binding protein (TBP)-associated factor 31 kD; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31 kD; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31 kDa; TAFII31; TAFII31L; TATA box binding protein associated factor 9 b; TATA-box binding protein associated factor 9 b; TBP-associated factor 9 L; TFIID-31; transcription associated factor TAFII31L; transcription initiation factor IID, 31 kD subunit; Transcription initiation factor TFIID subunit 9 B; Transcription initiation factor TFIID subunit 9-like; transcription-associated factor TAFII31L |
| Common Name | TAF9B |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction