Learn More
Human TLR3 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems AF1487
This item is not returnable.
View return policy
Description
TLR3 Polyclonal specifically detects TLR3 in Human samples. It is validated for Western Blot.
Specifications
| TLR3 | |
| Polyclonal | |
| Unconjugated | |
| O15455 | |
| TLR3 | |
| Mouse myeloma cell line NS0-derived recombinant human TLR3 Lys27-Ser711 Accession # O15455 | |
| 100 μg | |
| Primary | |
| Detects human TLR3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with rhTLR1, rhTLR2, rhTLR4 and rmTLR6 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CD283, CD283 antigen, toll-like receptor 3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7098 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction