Learn More
Invitrogen™ Human WDR77 (aa 14-104) Control Fragment Recombinant Protein
Recombinant Protein
Brand:  Invitrogen™ RP94078
Product Code. 30213137
  • 259.00 EUR / 100µL
Description
WDR77 is a component of the 20S PRMT5-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins. This modification targets Sm proteins to the survival of motor neurons (SMN) complex for assembly into small nuclear ribonucleoprotein core particles.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q9BQA1
Blocking Assay, Control
79084
100 μL
RUO
WDR77
Human
AREWNLPPNAPACMERQLEAARYRSDGALLLGASSLSGRCWAGSLWLFKDPCAAPNEGFCSAGVQTEAGVADLTWVGERGILVASDSGAVE
Liquid
≥5.0 mg/mL
1M urea, PBS with no preservative; pH 7.4
WDR77
-20° C, Avoid Freeze/Thaw Cycles
2610003I18Rik; 2610312E17Rik; Ac2-269; androgen receptor cofactor p44; C79984; HKMT1069; MEP50; MEP-50; Methylosome protein 50; MGC2722; Nbla10071; OTTHUMP00000013473; p44; p44/Mep50; RGD1310479; RP11-552M11.3; testis tissue sperm-binding protein Li 44a; WD repeat domain 77; WD repeat-containing protein 77; WD45; WDR77
Unconjugated
Recombinant
E. coli