Learn More
Proteintech™ HumanKine™ Recombinant Human HSA Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
Brand:  Proteintech™ HZ-3001-10000UG
 View more versions of this product
Product Code. 16871426
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
Specifications
213
10 x 1000 μg
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS.
Human embryonic kidney 293 (HEK293) human cell expression system
> 95%
60 to 65 kDa, monomer, glycosylated
Human
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Lyophilized