missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech HumanKine™ recombinant human IL-8 protein
Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
Description
IL-8 is a chemoattractant cytokine that signals through the CXCR1 and CXCR2 receptors to attract and activate neutrophils to sites of inflammation. It is secreted by multiple types of cells, including monocytes, neutrophils, tumor, epithelial, endothelial, and fibroblasts. IL-8 signaling activates downstream pathways that promote cell survival, proliferation, and cell invasiveness. As such, IL-8 has been linked to the promotion of angiogenesis, immunosuppressive cell recruitment, and EMT transition in several cancer types. Recent studies also indicate that IL-8 can serve as a marker for COVID-19 disease progression (PMID: 22235381, 18980965, 26548401, 33584733).
Specifications
Specifications
| Accession Number | P10145 |
| Formulation | 1 x PBS |
| Gene ID (Entrez) | 3576 |
| Molecular Weight (g/mol) | 12 kDa |
| Quantity | 100 μg |
| Source | HEK293 |
| Storage Requirements | Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.) |
| Endotoxin Concentration | <0.1 EU/μg |
| Specific Reactivity | Human |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction