missing translation for 'onlineSavingsMsg'
Learn More

Hyaluronic Acid, MP Biomedicals™

Product Code. 11450112
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in the study of activation of the MAPK ERK following Leishmania amazonensis, infection of macrophages. Hyaluronic Acid, MP Biomedicals is a high viscosity, high molecular mass polymer composed of repeating dimeric units of glucuronic acid and N-acety

A mucopolysaccharide composed of repeating dimeric units of glucuronic acid and N-acetyl glucosamine which forms the core of complex proteoglycan aggregates found in extracellular matrix.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9004-61-9
Molecular Formula C28H44N2O23
Quantity 50 mg
Synonym hyaluronic acid, dsstox_cid_26750, dsstox_rid_81875, dsstox_gsid_46750, 4xi-alpha-d-xylo-hexopyranuronosyl-1->3-3xi-2-acetylamino-2-deoxy-beta-d-ribo-hexopyranosyl-1->4-5xi-beta-d-xylo-hexopyranuronosyl-1->3-2-acetylamino-2-deoxy-d-glucopyranose
Solubility Information Dissolves at 10mg/mL in analytical grade water to give a clear, viscous solution, after soaking overnight at 5°C then stirring at room temperature for approximately 1 hour.
InChI Key KIUKXJAPPMFGSW-YXBJCWEESA-N
SMILES CC(=O)NC1C(C(C(OC1O)CO)O)OC2C(C(C(C(O2)C(=O)O)OC3C(C(C(C(O3)CO)O)OC4C(C(C(C(O4)C(=O)O)O)O)O)NC(=O)C)O)O
IUPAC Name (2S,4S,5R,6S)-6-[(2S,3R,5S,6R)-3-acetamido-2-[(3S,4R,5R,6R)-6-[(3R,4R,5S,6R)-3-acetamido-2,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-2-carboxy-4,5-dihydroxyoxan-3-yl]oxy-5-hydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Molecular Weight (g/mol) 776.651
PubChem CID 24728612
Formula Weight 50,000 to 8x106 (Lit.)
Physical Form Powder
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'