missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Newborn Bovine Serum, New Zealand Origin

Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA)

Brand:  Cytiva SH30560.02

 View more versions of this product

Product Code. 16665775

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Carefully collected, processed, and filtered in New Zealand to ensure safety against bovine diseases
TRUSTED_SUSTAINABILITY
Specifications

Specifications

<10 days
Newborn Calf Bovine Serum
Sequential Triple-filtered, 100 nm
≤ 25 EU/mL
HyClone™
New Zealand
1000 mL
None
≤ 25 mg/dL
European Agency for the Evaluation of Medicinal Products (EMEA) Conforming Tests
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers