Learn More
Cytiva HyClone™ Adult Bovine Serum, New Zealand Origin
Click to view available options
Quantity:
500 mL
1000 mL
3000 mL
Processing:
Irradiated
Non-Irradiated
Unit Size:
1 L
3 L
500mL
Specifications
Specifications
| Age | 6 to 8 years |
| Country of Origin | New Zealand |
| Product Type | Adult Bovine Serum |
| Quantity | 1000 mL |
| Sterilization Method | Triple-filtered |
| Processing | Non-Irradiated |
| Product Line | HyClone |
| Test Methods | Tested According to European Pharmacopoeia Protocols |