Learn More
Cytiva HyClone™ Adult Bovine Serum, New Zealand Origin
Product Code. 16655765
Click to view available options
Quantity:
500 mL
1000 mL
3000 mL
Processing:
Irradiated
Non-Irradiated
Unit Size:
1 L
3 L
500mL
Product Code. 16655765
Brand: Cytiva SH30528.05
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA), average age at time of collection is 6 to 8 years
TRUSTED_SUSTAINABILITY
Specifications
Age 6 to 8 years
Country of Origin New Zealand
Product Type Adult Bovine Serum
Quantity 3000 mL
Sterilization Method Triple-filtered
Processing Irradiated
Product Line HyClone
Test Methods Tested According to European Pharmacopoeia Protocols