Learn More
HYDRANAL™ - Solvent, Reagent for volumetric two-component KF titration (working medium), Honeywell Fluka™
Product Code. p-7072130 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 15674410
missing translation for 'mfr': Honeywell-Hydranal 348001L
Request Bulk or Custom Format
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Specifications
Chemical Name or Material HYDRANAL™-Solvent
CAS 67-56-1, 288-32-4, 7446-09-5
Density 0.89g/mL
Quantity 1 L
Flash Point 11°C (51.8°F)
MDL Number MFCD00147197
UN Number UN3286
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL-Solvent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H360D-May damage the unborn child.,May damage the unborn child.,May damage the unborn child.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.,Causes damage to organs.,Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : methanol, imidazole,sulphur dioxide