missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution, 0.01M, Chem-Lab

Product Code. 10592402
Click to view available options
Quantity:
1 L
Packaging:
Polyethylene
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as a laboratory reagent and acidifying agent. Hydrochloric Acid Solution, 0.01M, Chem-Lab contains 0.365g HCl per liter of water. It is used for pH control, titrations, the production of organic and inorganic compounds, and in the regeneration of ion exchange resins. ASTM D-4047

Hydrochloric acid is a corrosive, strong mineral acid with many industrial uses. It is a colorless, highly pungent solution of hydrogen chloride (HCl) in water. It is frequently used in pH buffer solutions, in the regeneration of ion exchange resins and in the production of chlorides and the synthesis of organic products.

Specifications

Color Colorless
CAS 7647-01-0, 7732-18-5
Composition 0.365g HCl per 1L H2O
Concentration 0.01mol/L
Density 1.00g/mL
Packaging Polyethylene
Normality 0.01N (±0.00002, 20°C)
Quantity 1 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.