missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid Solution, 0.5M, Chem-Lab
Click to view available options
Quantity:
1 L
Packaging:
Polyethylene
Unit Size:
1 L
Description
Hydrochloric acid is a corrosive, strong mineral acid with many industrial uses. It is a colorless, highly pungent solution of hydrogen chloride (HCl) in water. It is frequently used in pH buffer solutions, in the regeneration of ion exchange resins and in the production of chlorides and the synthesis of organic products.
Specifications
Specifications
| Color | Colorless |
| CAS | 7647-01-0, 7732-18-5 |
| Composition | 18.23g HCl per 1L H2O |
| Concentration | 0.5mol/L |
| Density | 1.01g/mL |
| Packaging | Polyethylene |
| Normality | 0.5N (±0.001, 20°C) |
| Quantity | 1 L |
| Molecular Formula | ClH |
| Physical Form | Liquid |
| Show More |
Safety and Handling
Product Identifier
- Hydrochloric acid 0.5 mol/l
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contains : HCl/H2O
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction