missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Solution, 37% AR, Chem-Lab

Product Code. 11979002
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as a laboratory reagent and acidifying agent. Hydrochloric Acid Solution, 37% AR, Chem-Lab is used for pH control, titrations, trace analysis, the production of organic and inorganic compounds, and in the regeneration of ion exchange resins. According to ISO, Ph. Eur. standards.

Hydrochloric acid is a corrosive, strong mineral acid with many industrial uses. It is a colorless, highly pungent solution of hydrogen chloride (HCl) in water. It is frequently used in pH buffer solutions, in the regeneration of ion exchange resins and in the production of chlorides and the synthesis of organic products.

Specifications

Appearance Colorless
pH <1
Color Colorless
CAS 7647-01-0, 7732-18-5
Composition Hydrochloric acid 37% a.r.
Melting Point -25°C
Boiling Point 50°C
Concentration 37+% HCl
Density 1.19g/mL
Residue after Ignition <0.0005%
Quantity 1 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Soluble in Water
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Percent Purity >37%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid 37% a.r.
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'