missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 3M (3N) Solution, Scharlab

Product Code. 12466781
Click to view available options
Quantity:
1 L
200 L
Packaging:
HDPE Bottle
Metal drum
Unit Size:
1 L
200 L
This item is not returnable. View return policy

Product Code. 12466781

Brand: Scharlab AC07381000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ready-to-use volumetric solution of accurate and reliable concentration for titration. Hydrochloric Acid, 3M (3N) Solution, Scharlab has a concentration that is exact to within the narrowest confidence interval for use as a reference solution in quantitative analysis.

  • Standard volumetric solution is ready to use
  • Certificate of Analysis details the factor, uncertainty level and methods used
  • Traceable to the SRM of NIST
  • Packaged in HDPE containers that fit into most titrators or in metal drums for bulk amounts

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless
Concentration 3mol/L
Density ∼1.06g/cm3
Packaging HDPE Bottle
Traceability to NIST Yes
Normality 3N
Quantity 1 L
Physical Form Liquid
UN Number 1789
Formula Weight 36.46g/mol
Concentration or Composition (by Analyte or Components) 1mL = 0.10938g HCl
Chemical Name or Material Hydrochloric Acid
Grade Checked potentiometricaly using Scharlau tris(hydroxymethyl)- aminomethane volumetric standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.