missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, TraceSELECT™, 30.0 to 32.0 %, Honeywell Fluka™
TraceSELECT™, for trace analysis,≥30%
78.67 EUR - 286.85 EUR
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
IUPAC Name | hydrogen chloride |
SMILES | Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15610880
|
Honeywell-Fluka
08256-100ML |
100 mL | Plastic Bottle |
103.49 EUR
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15640880
|
Honeywell-Fluka
08256-500ML |
500 mL | Plastic Bottle |
78.67 EUR
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15620880
|
Honeywell-Fluka
08256-1L |
1 L | Plastic Bottle |
133.74 EUR
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15630880
|
Honeywell-Fluka
08256-2.5L |
2.5 L | Plastic Bottle |
286.85 EUR
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |
Specifications
Colorless | |
-35°C | |
1.15g/mL at 20°C, 1.2g/mL at 25°C | |
ClH | |
MFCD00011324 MFCD00792839 | |
1098214 | |
Soluble in water | |
Cl | |
36.46 | |
CHEBI:17883 | |
30.0 to 32.0 % | |
Hydrochloric Acid |
7647-01-0 | |
42°C | |
3.23psi (21.1°C), 7.93psi (37.7°C) | |
Liquid | |
UN1789 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.46g/mol | |
TraceSELECT™ |
Safety and Handling
P261-P280-P305 + P351 + P338-P310
H290-H314-H335
missing translation for 'dotInformation' : 8/PGII
missing translation for 'einecsNumber' : 231-595-7
missing translation for 'rtecsNumber' : MW4025000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Hydrochloric acid
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.